Fiberlite F21-8 x 50y Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F21-8 x 50y Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Thermo Scientific
Fiberlite F21-8 x 50y Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Perform subcellular separations with the ergonomic Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor, a high-speed, carbon fiber rotor.
Have Questions?
Catalog NumberCapacity
096-0842758 x 50 mL
Catalog number 096-084275
Price (MYR)
43,076.00
EA
Add to cart
Capacity:
8 x 50 mL
Price (MYR)
43,076.00
EA
Add to cart
Perform subcellular separations with the ergonomic Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor, a high-speed, carbon fiber rotor.
Features:
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal
  • Corrosion- and fatigue-resistant construction secures the rotor's structural integrity for unequalled durability
  • Rotor is backed by a 15-year warranty
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionFiberlite F21-8 x 50y Fixed-Angle Rotor with Auto-Lock
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity8 x 50 mL
Max. RCF47,850 x g
Max. Speed20,000 rpm
Net Weight10.5 lbs. (4.8 kg)
For Use With (Equipment)Thermo Scientific Sorvall LYNX Superspeed Centrifuges
BiocontainmentYes
Unit SizeEA