Fiberlite F14-6 x 250y Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F14-6 x 250y Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Fiberlite F14-6 x 250y Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Improve ergonomics and performance with the Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-0620756 x 250 mL
Catalog number 096-062075
Price (MYR)
57,153.00
EA
Add to cart
Capacity:
6 x 250 mL
Price (MYR)
57,153.00
EA
Add to cart
Improve ergonomics and performance with the Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed-Angle Rotor, ideal for tissue homogenates or smaller volume yeast or bacterial culture separations. This lightweight rotor offers six-place capacity for bottles up to 250mL.
Features
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal
  • Corrosion and fatigue-resistant construction secures the rotor's structural integrity for unequaled durability
  • Backed by a 15-year warranty
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionFiberlite F14-6x250y Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity6 x 250 mL
Max. RCF30,240 x g
Max. Speed14,000 rpm
Radius (Metric)138 mm
Radius (Metric) Min.37 mm
K Factor1,699
Max. Tube Length135 mm
Max. Tube Diameter62 mm
Net Weight17.5 lb. (8.0 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle23°
BiocontainmentYes
Unit SizeEA