Fiberlite F9-6 x 1000 LEX Fixed Angle Rotor for LYNX 6000 Superspeed Centrifuge
Fiberlite F9-6 x 1000 LEX Fixed Angle Rotor for LYNX 6000 Superspeed Centrifuge
Actual product may vary
Fiberlite F9-6 x 1000 LEX Fixed Angle Rotor for LYNX 6000 Superspeed Centrifuge
Shorten set-up time while providing peace of mind that the rotor is secure with Thermo Scientific™ Fiberlite™ F9-6 x 1000mL LEX Fixed Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-0610756 x 1000 mL
Catalog number 096-061075
Price (MYR)
133,244.00
EA
Add to cart
Capacity:
6 x 1000 mL
Price (MYR)
133,244.00
EA
Add to cart
Shorten set-up time while providing peace of mind that the rotor is secure with Thermo Scientific™ Fiberlite™ F9-6 x 1000mL LEX Fixed Angle Rotor, designed for bioprocessing (up to 6L per run) and offering the flexibility of adapters for labware from 1.5 to 500mL.
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Improved ergonomics and performance:
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal.
  • Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability.
  • Rotor is backed by a 15-year warranty.
Specifications
DescriptionFiberlite F9-6x1000 LEX Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity6 x 1000 mL
Max. RCF17,568 x g
Max. Speed9,000 rpm
Radius (Metric)194 mm
Radius (Metric) Min.65 mm
K Factor3,415
Max. Tube Length195 mm
Max. Tube Diameter98 mm
Net Weight34.5 lb. (15.7 kg)
For Use With (Equipment)Sorvall LYNX 6000 Superspeed Centrifuge
Angle20°
BiocontainmentYes
Unit SizeEA