Fiberlite F10-4 x 1000 LEX Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F10-4 x 1000 LEX Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Fiberlite F10-4 x 1000 LEX Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Process lower-volume samples with this innovative fixed angle rotor which offers rotor support adapters for tubes from 1.5 to 50mL.
Have Questions?
Catalog NumberCapacity
096-041075
also known as 096-041075EX
4 x 1000 mL
Catalog number 096-041075
also known as 096-041075EX
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
4 x 1000 mL
Process lower-volume samples with the innovative Thermo Scientific™ Fiberlite™ F10-4 x 1000 LEX Fixed Angle Rotor, which offers rotor support adapters for tubes from 1.5 to 50mL. A lightweight carbon fiber construction allows for a unique rotor design not possible with metal, while corrosion and fatigue-resistance secures the rotor's structural integrity. Backed by a 15-year warranty.
Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionFiberlite F10-4x1000 LEX Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity4 x 1000 mL
Max. RCF20,584 x g
Max. Speed10,500 rpm
Radius (Metric)167 mm
Radius (Metric) Min.26 mm
K Factor4,358
Max. Tube Length195 mm
Max. Tube Diameter98 mm
Net Weight24.0 lb. (10.9 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle20°
BiocontainmentYes
Unit SizeEA