Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Have Questions?
Change view
Catalog Number
Type
09.4003
Pretreatment Filter
09.4000
Pretreatment Cartridge
06.5204
Filter with Stabilizer
06.5452
Stabilizing Cartridge
50134022
Filter
5 Options
Catalog number 09.4003
Price (MYR)
1,231.00
EA
Add to cart
Type:
Pretreatment Filter
Price (MYR)
1,231.00
EA
Add to cart
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
For Use With (Equipment)Smart2Pure, LabTower RO, LabTower TII