Smart2Pure 12 Water Purification System RO Membrane with Integrated Pretreatment
Product Image
Smart2Pure 12 Water Purification System RO Membrane with Integrated Pretreatment
Have Questions?
Catalog number 09.2012
Price (MYR)
3,956.00
EA
Add to cart
Price (MYR)
3,956.00
EA
Add to cart
  • RO membrane with integrated pretreatment
  • For use with Smart2Pure water purification system
  • 12L/hr flow rate
Specifications
Cartridge TypeReverse Osmosis Membrane
DescriptionReverse osmosis with integrated pretreatment
Flow Rate12 L/hr.
Product Water TypeType 3
TypeReplacement part for Smart2Pure Water Purification System
For Use With (Application)AA, IC, ICP, standard buffer
RO Flow Rate12 L/hr.
Unit SizeEA