Specifications
Quantity5/Pk.
Length (Metric)7.85 cm
Packing MaterialQuartz Wool
Product TypeSplit/Splitless Fast Focus Liner
Diameter (Metric) Inner0.23 cm
Diameter (Metric) Outer0.63 cm
For Use With (Equipment)Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
DescriptionUltrapure polisher cartridge
For Use With (Equipment)GenPure GenPure Pro, GenPure xCAD, LabTower EDI
TypeCartridge, Ultrapure Polisher
Unit SizeEA