Point of Use Filter Inserts
Point of Use Filter Inserts
Actual product may vary
Point of Use Filter Inserts
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFlow RateFor Use With (Equipment)
06.5201
also known as T06.5201
Prefilter 5µm+activated carbon (10“)750 L/hr. (at D p = 0.2 bar)LabTower EDI, LabTower RO
06.5101
also known as T06.5101
Prefilter 1µm (10“)1000 L/hr. (at D p = 0.2 bar)Smart2Pure
Catalog number 06.5201
also known as T06.5201
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Prefilter 5µm+activated carbon (10“)
Flow Rate:
750 L/hr. (at D p = 0.2 bar)
For Use With (Equipment):
LabTower EDI, LabTower RO
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters. Thermo Scientific™ Point of Use Filter Inserts include 10- and 20-inch pre-filters, final filters and sterile filters for Point of Use systems.
Pre-filters 1μm, 5μm
  • Protect installations and systems from undissolved substances in water
Final filters
  • Protect systems from undissolved substances in water
  • Usually used for final filtration
Sterile filters
  • For final filtration and delivery of organic-free water
Specifications
Cartridge Size10 in.
DescriptionPrefilter 5µm+activated carbon (10“)
For Use With (Equipment)LabTower EDI, LabTower RO
TypePrefilter Insert
Flow Rate750 L/hr. (at D p = 0.2 bar)
Pore Size (Metric)5 μm
Unit SizeEA