Fiberlite 250 mL Bottle
Fiberlite 250 mL Bottle
Actual product may vary
Fiberlite 250 mL Bottle
Thermo Scientific™ 250mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottle will be available for Thermo Scientific™ Sorvall™ LYNX Superspeed and General Purpose Centrifuges
Have Questions?
Catalog NumberMaterial
010-1495Polypropylene
Catalog number 010-1495
Price (MYR)
1,381.00
PK
Add to cart
Material:
Polypropylene
Price (MYR)
1,381.00
PK
Add to cart
Thermo Scientific™ 250mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottle will be available for Thermo Scientific™ Sorvall™ LYNX Superspeed and General Purpose Centrifuges.
Features
  • PPCO & PC
  • Increased cycle life compared to previous Thermo Scientific bottles - 100 cycle life (caps + bottles)
  • Certifications USP Class VI - Non-Cytotoxic
  • Max speeds match rotors
  • Ergonomic speed handle design allows much easier bottle handling and removal
Specifications
Capacity (Metric)250 mL
MaterialPolypropylene
Product LineFiberlite
Unit SizePK
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.