Centrifuge Polycarbonate Bottle
Centrifuge Polycarbonate Bottle
Actual product may vary
Centrifuge Polycarbonate Bottle
39mL Centrifuge Polycarbonate Bottle
Have Questions?
Catalog number 010-1370
Price (MYR)
1,792.00
EA
Add to cart
Price (MYR)
1,792.00
EA
Add to cart
39mL Centrifuge Polycarbonate Bottle
  • 39 mL
  • Centrifuge Polycarbonate Bottle
  • Fill Volume 26.3 mL
  • Diameter 25 mm
  • Length 89 mm
  • Supplied Without Closure
  • Packet of 6
  • For Fiberlite F37L-8x100 Rotor
Specifications
TypePolycarbonate Bottle
MaterialPolycarbonate
For Use With (Equipment)Ultracentrifuge Fiberlite F37L-8 x 100 Rotor
Capacity26.3 mL
Max. Speed30,000 rpm
Tube Size39 mL
Diameter (Metric) Outside25 mm
Length (Metric)89 mm
No. per Pack6
Unit SizeEA