Check Order Status
15 mL Nunc Conical Tube or Filtration Devices Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
15 mL Nunc Conical Tube or Filtration Devices Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Accommodate 15mL conical or filtration devices in Thermo Scientific™ Fiberlite™ conical tube rotors.
Have Questions?
Catalog number
010-1340
Price (MYR)
-
Accommodate 15mL Nunc conical tube or filtration devices in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
Type
Filtration Devices Adapter
Tube Type
Conical
Capacity
15 mL
For Use With (Equipment)
Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description
15mL Nunc Conical Tube or Filtration Devices Adapter
Tube Size
15 mL