10 mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Product Image
10 mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Accommodate 10mL Oak Ridge or 12mL round tubes in Thermo Scientific™ Fiberlite™ conical tube rotors.
Have Questions?
Catalog number 010-1311
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Accommodate 10mL Nalgene Oak Ridge or 12mL round tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
TypeRidge Tube Adapter
Tube Type10 mL Nalgene Oak Ridge Tube, 12 mL Round Tube
Capacity10, 12 mL
For Use With (Equipment)Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description10mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapter
Unit SizePK