Check Order Status
PTFE Stopper, Self Releasing
PTFE Stopper, Self Releasing
Fisherbrand™ PTFE Stopper, Self Releasing fit standard taper ground glass joints with sealing rings and self releasing ring for ease of removal.
Have Questions?
Catalog Number
Standard Taper Joint
Height (Metric)
010-1119
19/38
62 mm
Catalog number
010-1119
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Standard Taper Joint:
19/38
Height (Metric):
62 mm
Total Chemical Resistance
High Purity
Temperature Range -200°C to +280°C
Non contaminating
Easy to clean
Specifications
Description
Fisherbrand™ PTFE Stopper, Self Releasing, Full Length A Type
For Use With (Equipment)
Standard Taper Ground Glass Joints
Height (Metric)
62 mm
Quantity
1
Standard Taper Joint
19/38
Temperature Range (Metric)
-200°C to +280°C
Material
PTFE