Check Order Status
16 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
16 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Accommodate 16mL Oak Ridge tubes in Thermo Scientific™ Fiberlite™ conical tube rotors.
Have Questions?
Catalog number
010-0376
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Accommodate 16mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
Type
Ridge Tube Adapter
Tube Type
16 mL Nalgene Oak Ridge Tube
Capacity
16 mL
For Use With (Equipment)
Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description
16mL Nalgene Oak Ridge Tube Adapter