Vanadium(V) oxide, 98+%
Vanadium(V) oxide, 98+%
Vanadium(V) oxide, 98+%
Vanadium(V) oxide, 98+%
Thermo Scientific Chemicals
Vanadium(V) oxide, 98+%
CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA207995000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point1750.0°C
Chemical Name or MaterialVanadium(V) oxide
Melting Point690.0°C
Name Note98+%
CAS1314-62-1