Potassium selenocyanate, 99%
Potassium selenocyanate, 99%
Potassium selenocyanate, 99%
Potassium selenocyanate, 99%
Thermo Scientific Chemicals
Potassium selenocyanate, 99%
CAS: 3425-46-5 | CKNSe | 144.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA201980500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium selenocyanate
Melting Point152.0°C to 158.0°C
CAS3425-46-5
ColorOrange to Red
Health Hazard 1GHS Signal Word: Danger