5-Fluorosalicylic acid, 97%
5-Fluorosalicylic acid, 97%
5-Fluorosalicylic acid, 97%
5-Fluorosalicylic acid, 97%
5-Fluorosalicylic acid, 97%
CAS: 345-16-4 | C7H5FO3 | 156.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number ALFB21751.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material5-Fluorosalicylic acid
CAS345-16-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Wilfried Rozhon; Elena Petutschnig; Michael Wrzaczek; Claudia Jonak. Quantification of free and total salicylic acid in plants by solid-phase extraction and isocratic high-performance anion-exchange chromatography.Analytical and Bioanalytical Chemistry.200538(7), 1620-1627.
  2. T K Christopoulos; E P Diamandis. Enzymatically amplified time-resolved fluorescence immunoassay with terbium chelates.Analytical Chemistry.199264(4), 342-346.