2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
2-Naphthoxyacetic acid, 97%
Thermo Scientific Chemicals
2-Naphthoxyacetic acid, 97%
CAS: 120-23-0 | C12H10O3 | 202.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA15412.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Naphthoxyacetic acid
CAS120-23-0
Health Hazard 1H302-H315-H319
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Notes
Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Tsilikochrisos, G.; Tsaniklidis, G.; Delis, C.; Nikoloudakis, N.; Aivalakis, G. Glutamate dehydrogenase is differentially regulated in seeded and parthenocarpic tomato fruits during crop development and postharvest storage. Sci. Hortic. 2015, 181, 34-42.
  2. Song, X. Y.; Chen, J.; Shi, Y. P. Electromembrane extraction based on carbon nanotubes reinforced hollow fiber for the determination of plant hormones. New J. Chem. 2015, 39 (12), 9191-9199.