6-Benzyladenine, 99%
6-Benzyladenine, 99%
6-Benzyladenine, 99%
6-Benzyladenine, 99%
Thermo Scientific Chemicals
6-Benzyladenine, 99%
CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA14678.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material6-Benzyladenine
CAS1214-39-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Notes
Incompatible with strong oxidizing agent.
RUO – Research Use Only
General References:
  1. Bound, S. A. Optimising crop load and fruit quality of ‘Packham’s Triumph’ pear with ammonium thiosulfate, ethephon and 6-benzyladenine. Sci. Hortic. 2015, 192, 187-196.
  2. Kaviani, B.; Hashemabadi, D.; Khodabakhsh, H.; Onsinejad, R.; Ansari, M. H.; Haghighat, N. Micropropagation of Begonia rex Putz. by 6-benzyladenine and alpha-naphthalene acetic acid. Int. J. Biosci. 2015, 6 (5), 8-15.