Phenyltrimethoxysilane, 97%
Phenyltrimethoxysilane, 97%
Phenyltrimethoxysilane, 97%
Phenyltrimethoxysilane, 97%
Thermo Scientific Chemicals
Phenyltrimethoxysilane, 97%
CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13827.22100 g
Catalog number ALFA13827.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point210°C to 212°C
Chemical Name or MaterialPhenyltrimethoxysilane
Melting Point-25°C
CAS2996-92-1
Health Hazard 1H226-H302-H373
View more
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Touria Jermouni; Monique Smaihi and Nadine Hovnanian. Hydrolysis and initial polycondensation of phenyltrimethoxysilane and diphenyldimethoxysilane.J. Mater. Chem.1995, 5 1203-1208.
  2. Yung-Chih Chen; Hsueh-Chen Lin; Yu-Der Lee. The Effects of Phenyltrimethoxysilane Coupling Agents on the Properties of PTFE/Silica Composites.Journal of Polymer Research.2004, 11 1-7
  3. Aryltrimethoxysilanes undergo Pd-catalyzed cross-coupling reactions with e.g. aryl iodides in the presence of TBAF, via a postulated hypervalent siloxane intermediate: J. Org. Chem., 64, 1684 (1999):