p-Tolyl isothiocyanate, 97%
p-Tolyl isothiocyanate, 97%
p-Tolyl isothiocyanate, 97%
p-Tolyl isothiocyanate, 97%
Thermo Scientific Chemicals
p-Tolyl isothiocyanate, 97%
CAS: 622-59-3 | C8H7NS | 149.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA11499.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Tolyl isothiocyanate
CAS622-59-3
Health Hazard 1H301+H311+H331-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
View more
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.
Solubility
Decompose in water.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
RUO – Research Use Only
General References:
  1. Thorfinnur Gunnlaugsson; Paul E Kruger; Paul Jensen; Juliann Tierney; Haslin Dato Paduka Ali; Gillian M Hussey. Colorimetric naked eye sensing of anions in aqueous solution. Journal of Organic Chemistry.2005, 70 (26), 10875-10878.
  2. D H Swenson; K el-Bayoumy; G H Shuie; S S Hecht; E Fiala; F F Kadlubar; J P Freeman. Synthesis of N-(purin-8-yl)arylamines. Chemical Research in Toxicology.1993, 6 (4), 480-485.