Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits
Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits
Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits
Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits
Thermo Scientific Chemicals
Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits
CAS: 61789-51-3 | C20H34CoO4 | 397.42 g/mol
Have Questions?
Catalog NumberQuantity
ALF040387.36500 g
Catalog number ALF040387.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt naphthenate
CAS61789-51-3
Health Hazard 1H226-H302-H304-H317-H336-H350-H372
Health Hazard 2GHS H Statement
H304-H351-H226-H317
May be fatal if swallowed and enters airways.
Suspected of causing cancer.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P330-P331-P333+P313-P363-P370+P378q-P501c
View more
Cobalt naphthenate is widely used in polyester resins and paint driers. It is used as a curing accelerator, cross linker catalyst for unsaturated polyester resins. It is also used in the production of adhesives, varnishes and waterproofing agents for textiles. Further, it is used as corrosion inhibitors, lubricants, and fuel additives. It plays an important role as oil drying agents.