LabServ 9" PF Nitrile Glove, S, 100/Bx
Product Image
LabServ 9" PF Nitrile Glove, S, 100/Bx
Have Questions?
Catalog number LVG420948S
Price (MYR)
20.00
Special offer
Ends: 31-Dec-2025
24.00
Save 4.00 (17%)
BX
Add to cart
Price (MYR)
20.00
Special offer
Ends: 31-Dec-2025
24.00
Save 4.00 (17%)
BX
Add to cart
Specifications
Unit SizeBX