Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Thermo Scientific Chemicals
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
CAS: 7789-78-8 | CaH2 | 42.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA199905000
Price (MYR)
2,219.00
Bottle
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,219.00
Bottle
Specifications
Chemical Name or MaterialCalcium hydride
Melting Point675.0°C
Name Noteextra pure, 0-2 mm grain size
CAS7789-78-8
Health Hazard 1GHS Signal Word: Danger